Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018837.1 | NP_001018238.1 | 6 | 731 | ||
NM_006118.3 | NP_006109.2 | 6 | 875 | |||
GenBank mRNA | AK290626.1 | - | 6 | 797 | ||
AK300676.1 | - | 6 | 853 | |||
AK310000.1 | - | 6 | 718 | |||
AK311450.1 | - | 3 | 194 | |||
AK313291.1 | - | 6 | 806 | |||
BC005240.1 | - | 6 | 927 | 3931805 | ||
BC014314.1 | - | 6 | 796 | 3931852 | ||
BC015209.1 | - | 6 | 768 | 3883802 | ||
BC016730.1 | - | 6 | 813 | 4082810 | ||
EU190982.1 | - | 5 | 984 | |||
EU190983.1 | - | 6 | 792 | |||
KU178523.1 | - | 6 | 714 | |||
KU178524.1 | - | 6 | 570 | |||
KU178525.1 | - | 4 | 263 | |||
U68566.1 | - | 6 | 875 |