Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TACC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006342.2 | NP_006333.1 | 8 | 1819 | ||
XM_005247929.2 | XP_005247986.1 | 8 | 1817 | |||
XM_005247930.2 | XP_005247987.1 | 7 | 846 | |||
XM_011513386.2 | XP_011511688.1 | 8 | 1817 | |||
XM_017007653.1 | XP_016863142.1 | 8 | 1871 | |||
GenBank mRNA | AF093543.1 | - | 8 | 1772 | ||
AF479620.1 | - | 7 | 685 | |||
AJ243997.1 | - | 8 | 1764 | |||
AK302463.1 | - | 8 | 1790 | |||
AK304579.1 | - | 7 | 1548 | |||
AK310660.1 | - | 7 | 1542 | |||
BC106071.1 | - | 8 | 1780 | 6148176 | ||
BC111771.1 | - | 8 | 1732 | 40034847 |