Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MERTK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006343.2 | NP_006334.2 | 6 | 988 | ||
XM_005263565.4 | XP_005263622.1 | 6 | 1123 | |||
XM_005263568.4 | XP_005263625.1 | 6 | 1123 | |||
XM_011510490.2 | XP_011508792.1 | 6 | 1023 | |||
XM_017003164.1 | XP_016858653.1 | 6 | 993 | |||
XM_017003165.1 | XP_016858654.1 | 6 | 1040 | |||
GenBank mRNA | AK316584.1 | - | 6 | 995 | ||
AL705756.1 | - | 5 | 448 | |||
BC114917.1 | - | 6 | 702 | 40068588 | ||
BC114918.1 | - | 6 | 702 | 40012335 | ||
CA454601.1 | - | 4 | 378 | |||
U08023.1 | - | 6 | 1003 |