Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC30A9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006345.3 | NP_006336.3 | 10 | 1011 | ||
XM_017007654.1 | XP_016863143.1 | 10 | 1045 | |||
GenBank mRNA | AF006621.2 | - | 10 | 985 | ||
AK225470.1 | - | 10 | 948 | |||
AK299917.1 | - | 11 | 1105 | |||
AK312838.1 | - | 10 | 1011 | |||
AY319413.1 | - | 11 | 890 | |||
AY594282.1 | - | 10 | 865 | |||
BC000240.1 | - | 10 | 973 | |||
BC007732.2 | - | 10 | 972 | 4109702 | ||
BC016949.1 | - | 10 | 990 | 3850521 | ||
BC022981.1 | - | 6 | 402 | |||
KU178526.1 | - | 10 | 865 | |||
KU178527.1 | - | 10 | 865 |