Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIBF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006346.2 | NP_006337.2 | 5 | 902 | ||
XM_011534881.2 | XP_011533183.1 | 5 | 930 | |||
XM_011534882.2 | XP_011533184.1 | 5 | 930 | |||
XM_011534884.2 | XP_011533186.1 | 5 | 930 | |||
XM_011534885.2 | XP_011533187.1 | 4 | 557 | |||
XM_011534886.2 | XP_011533188.1 | 5 | 930 | |||
XM_017020350.1 | XP_016875839.1 | 4 | 556 | |||
XM_017020351.1 | XP_016875840.1 | 5 | 930 | |||
XM_017020352.1 | XP_016875841.1 | 5 | 856 | |||
XM_017020353.1 | XP_016875842.1 | 5 | 853 | |||
XM_017020354.1 | XP_016875843.1 | 5 | 853 | |||
GenBank mRNA | AF330046.1 | - | 5 | 944 | ||
AK309828.1 | - | 5 | 859 | |||
BC006009.1 | - | 5 | 855 | |||
BC008432.1 | - | 5 | 855 | |||
BC041806.1 | - | 5 | 854 | 5269898 | ||
BC051911.1 | - | 5 | 832 | 6140514 |