Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COG5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161520.1 | NP_001154992.1 | 9 | 1549 | ||
NM_006348.3 | NP_006339.3 | 9 | 1549 | |||
NM_181733.2 | NP_859422.2 | 9 | 1549 | |||
GenBank mRNA | AF058718.1 | - | 9 | 1075 | ||
AK314270.1 | - | 9 | 1547 | |||
BC068540.1 | - | 9 | 1296 | 4374289 |