Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC23B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172745.1 | NP_001166216.1 | 9 | 1380 | ||
NM_001172746.1 | NP_001166217.1 | 9 | 1416 | |||
NM_006363.4 | NP_006354.2 | 9 | 1474 | |||
NM_032985.4 | NP_116780.1 | 9 | 1492 | |||
NM_032986.3 | NP_116781.1 | 9 | 1171 | |||
XM_017027593.1 | XP_016883082.1 | 9 | 1368 | |||
GenBank mRNA | AK296270.1 | - | 9 | 1092 | ||
AK299171.1 | - | 9 | 1093 | |||
BC001575.1 | - | 9 | 1087 | |||
BC005032.1 | - | 9 | 1134 | 3607346 | ||
BC005404.1 | - | 9 | 1281 | 3460841 | ||
BC095404.1 | - | 9 | 1128 | 30339834 | ||
X97065.1 | - | 9 | 1136 |