Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006369.4 | NP_006360.3 | 8 | 2081 | ||
XM_017000074.1 | XP_016855563.1 | 8 | 2107 | |||
GenBank mRNA | AF370430.1 | - | 8 | 1981 | ||
AK125827.1 | - | 7 | 3358 | |||
AK291283.1 | - | 8 | 2064 | |||
AK296436.1 | - | 8 | 1900 | |||
AK297596.1 | - | 8 | 2095 | |||
BC004953.1 | - | 5 | 1317 | |||
BC101522.1 | - | 8 | 2071 | 8069028 | ||
BC101524.1 | - | 8 | 2071 | 8069030 | ||
BX537366.1 | - | 8 | 1991 | |||
X86018.1 | - | 5 | 1293 |