Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UNC13B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006377.3 | NP_006368.3 | 35 | 4343 | ||
XM_011517681.2 | XP_011515983.1 | 28 | 9507 | |||
XM_011517682.2 | XP_011515984.1 | 28 | 9505 | |||
XM_011517683.2 | XP_011515985.1 | 28 | 9503 | |||
XM_011517684.2 | XP_011515986.1 | 35 | 4361 | |||
XM_011517685.2 | XP_011515987.1 | 35 | 4358 | |||
XM_011517686.2 | XP_011515988.1 | 26 | 3535 | |||
XM_017014191.1 | XP_016869680.1 | 26 | 3396 | |||
XM_017014192.1 | XP_016869681.1 | 28 | 3313 | |||
GenBank mRNA | AB209998.1 | - | 35 | 4326 | ||
AF020202.1 | - | 35 | 4275 | |||
BC111781.1 | - | 35 | 4231 | 40037278 | ||
BC111800.1 | - | 37 | 4260 | 40037280 |