Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CIB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271888.1 | NP_001258817.1 | 3 | 608 | ||
NM_001271889.1 | NP_001258818.1 | 2 | 496 | |||
NM_001301224.1 | NP_001288153.1 | 3 | 658 | |||
NM_006383.3 | NP_006374.1 | 4 | 643 | |||
NR_125435.1 | 3 | 531 | ||||
XM_005254126.3 | XP_005254183.1 | 4 | 1084 | |||
XM_006720374.2 | XP_006720437.1 | 5 | 468 | |||
XM_011521161.2 | XP_011519463.1 | 4 | 352 | |||
GenBank mRNA | AB012955.1 | - | 4 | 329 | ||
AI817087.1 | - | 3 | 267 | |||
AI857912.1 | - | 3 | 358 | |||
AK293167.1 | - | 4 | 557 | |||
BC033108.1 | - | 3 | 608 | 4841632 | ||
BC047381.1 | - | 4 | 571 | 5287072 | ||
BC057292.1 | - | 4 | 571 | |||
BE042882.1 | - | 4 | 379 | |||
BM908035.1 | - | 270 | ||||
BQ681471.1 | - | 669 | ||||
BT019962.1 | - | 4 | 323 | |||
BU556614.1 | - | 2 | 394 | |||
CN371150.1 | - | 2 | 280 |