Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CIB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277764.1 | NP_001264693.1 | 4 | 597 | ||
NM_006384.3 | NP_006375.2 | 4 | 477 | |||
NR_102427.1 | 4 | 501 | ||||
NR_102428.1 | 4 | 367 | ||||
XM_006720375.1 | XP_006720438.1 | 4 | 472 | |||
GenBank mRNA | AB451276.1 | - | 4 | 315 | ||
AB451406.1 | - | 4 | 315 | |||
BC000846.2 | - | 4 | 357 | 3457844 | ||
BG231771.1 | - | 4 | 504 | |||
BG759961.1 | - | 4 | 367 | |||
BI520476.1 | - | 4 | 501 | |||
CD622547.1 | - | 4 | 484 | |||
CR456955.1 | - | 4 | 315 | |||
JQ246073.1 | - | 4 | 435 | |||
U82226.1 | - | 4 | 361 | |||
U83236.1 | - | 4 | 383 | |||
U85611.1 | - | 4 | 381 |