Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CIB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277764.1 | NP_001264693.1 | 5 | 714 | ||
NM_006384.3 | NP_006375.2 | 5 | 594 | |||
NR_102427.1 | 5 | 618 | ||||
NR_102428.1 | 5 | 484 | ||||
XM_006720375.1 | XP_006720438.1 | 5 | 589 | |||
GenBank mRNA | AA769426.1 | - | 3 | 398 | ||
AB451276.1 | - | 5 | 432 | |||
AB451406.1 | - | 5 | 432 | |||
BC000846.2 | - | 5 | 474 | 3457844 | ||
BG231771.1 | - | 3 | 387 | |||
BG759961.1 | - | 5 | 484 | |||
BI520476.1 | - | 5 | 618 | |||
CR456955.1 | - | 5 | 432 | |||
JQ246073.1 | - | 5 | 552 | |||
U82226.1 | - | 5 | 478 | |||
U83236.1 | - | 5 | 500 | |||
U85611.1 | - | 5 | 498 |