Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098504.1 | NP_001091974.1 | 13 | 1917 | ||
NM_006386.4 | NP_006377.2 | 13 | 1911 | |||
GenBank mRNA | AB209595.1 | - | 13 | 1873 | ||
AF131750.1 | - | 2 | 1213 | |||
AK024985.1 | - | 13 | 1879 | |||
AK074224.1 | - | 13 | 1879 | |||
AL713763.1 | - | 12 | 5805 | |||
BC000595.2 | - | 13 | 1858 | 3345982 | ||
CR456432.1 | - | 13 | 1607 | |||
U59321.1 | - | 13 | 1571 |