Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IPO7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006391.2 | NP_006382.1 | 17 | 2039 | ||
GenBank mRNA | AF098799.1 | - | 17 | 2004 | ||
AK074908.1 | - | 10 | 1070 | |||
AK312885.1 | - | 17 | 2010 | |||
BC114929.1 | - | 17 | 1965 | 40036258 |