Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNASEH2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006397.2 | NP_006388.2 | 3, 4 | 389 | ||
XM_006722619.2 | XP_006722682.1 | 4, 5 | 574 | |||
GenBank mRNA | AK315327.1 | - | 3, 4 | 419 | ||
BC011748.2 | - | 3, 4 | 380 | 3160621 | ||
BG825340.1 | - | 3, 4 | 393 | |||
CR456841.1 | - | 3, 4 | 329 | |||
Z97029.1 | - | 3, 4 | 389 |