Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAMTOR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006402.2 | NP_006393.2 | 3 | 434 | ||
GenBank mRNA | AA443738.1 | - | 2 | 424 | ||
AF029890.1 | - | 3 | 185 | |||
AK054769.1 | - | 2 | 1410 | |||
AY623819.1 | - | 3 | 130 | |||
BC062619.1 | - | 3 | 452 | 5190406 | ||
BX092018.1 | - | 3 | 450 | |||
CR456866.1 | - | 3 | 130 | |||
CR542130.1 | - | 3 | 130 |
Gene Symbol: LAMTOR5P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY669118.1 | - | 1 | 124 |