Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PROCR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006404.4 | NP_006395.2 | 3 | 746 | ||
XM_011528496.1 | XP_011526798.1 | 3 | 589 | |||
GenBank mRNA | AK314887.1 | - | 3 | 712 | ||
BC014451.1 | - | 3 | 610 | 4907433 | ||
BQ939781.1 | - | 1 | 78 | |||
CR456948.1 | - | 3 | 528 | |||
L35545.1 | - | 3 | 552 | |||
X89079.1 | - | 3 | 557 |