Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC34A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177998.1 | NP_001171469.1 | 13 | 1639 | ||
NM_001177999.1 | NP_001171470.1 | 13 | 1662 | |||
NM_006424.2 | NP_006415.2 | 13 | 1642 | |||
GenBank mRNA | AF111856.1 | - | 13 | 1624 | ||
AF146796.1 | - | 13 | 1593 | |||
AK075015.1 | - | 7 | 768 | |||
AK075046.1 | - | 12, 13 | 1427 | |||
AK292675.1 | - | 13 | 1630 | |||
AK314514.1 | - | 13 | 1651 | |||
BC142704.1 | - | 13 | 1587 | 40148805 | ||
BC146666.1 | - | 13 | 1587 | 40148124 | ||
KM822236.1 | - | 12 | 1558 |