Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006432.3 | NP_006423.1 | 2 | 207 | ||
XM_017020930.1 | XP_016876419.1 | 2 | 301 | |||
GenBank mRNA | AK222474.1 | - | 2 | 127 | ||
AK298975.1 | - | 2 | 107 | |||
AK300879.1 | - | 2 | 134 | |||
AK311931.1 | - | 2 | 168 | |||
BC002532.2 | - | 2 | 206 | 3140870 | ||
X67698.1 | - | 2 | 110 |