Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042550.1 | NP_001036015.1 | 22 | 3410 | ||
NM_001042551.1 | NP_001036016.1 | 22 | 3348 | |||
NM_001265602.1 | NP_001252531.1 | 22 | 3364 | |||
NM_006444.2 | NP_006435.2 | 22 | 3339 | |||
XM_006716933.3 | XP_006716996.1 | 22 | 3261 | |||
XM_011518148.2 | XP_011516450.1 | 22 | 3224 | |||
XM_011518149.2 | XP_011516451.1 | 22 | 3266 | |||
XM_011518150.2 | XP_011516452.1 | 21 | 3128 | |||
XM_011518151.2 | XP_011516453.1 | 21 | 3106 | |||
XM_011518152.2 | XP_011516454.1 | 22 | 3290 | |||
XM_011518153.1 | XP_011516455.1 | 22 | 3290 | |||
XM_017014206.1 | XP_016869695.1 | 22 | 3410 | |||
XM_017014207.1 | XP_016869696.1 | 22 | 3312 | |||
XM_017014208.1 | XP_016869697.1 | 22 | 3352 | |||
XM_017014209.1 | XP_016869698.1 | 21 | 3239 | |||
XM_017014210.1 | XP_016869699.1 | 21 | 3129 | |||
XM_017014211.1 | XP_016869700.1 | 21 | 3218 | |||
XM_017014212.1 | XP_016869701.1 | 21 | 3085 | |||
GenBank mRNA | AF092563.1 | - | 22 | 3274 | ||
AF113673.1 | - | 3 | 344 | |||
AK000955.1 | - | 11 | 1581 | |||
AK001485.1 | - | 22 | 3396 | |||
AK292599.1 | - | 22 | 3287 | |||
AL833191.1 | - | 22 | 3349 | |||
BC130385.1 | - | 22 | 3194 | 40146416 | ||
BC144163.1 | - | 22 | 3210 | 9052683 |