Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AHSA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012111.2 | NP_036243.1 | 1, 2 | 247 | ||
GenBank mRNA | AF164791.1 | - | 1, 2 | 153 | ||
AJ243310.1 | - | 1, 2 | 218 | |||
AK300766.1 | - | 1, 2 | 245 | |||
AK313824.1 | - | 1, 2 | 181 | |||
BC000321.2 | - | 1, 2 | 195 | 2821759 | ||
BG432594.1 | - | 1, 2 | 248 |