Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AHSA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321441.1 | NP_001308370.1 | 2 | 252 | ||
NM_012111.2 | NP_036243.1 | 2 | 386 | |||
GenBank mRNA | AF161440.1 | - | 1 | 30 | ||
AF164791.1 | - | 2 | 292 | |||
AJ243310.1 | - | 2 | 357 | |||
AK300766.1 | - | 2 | 384 | |||
AK313824.1 | - | 2 | 320 | |||
BC000321.2 | - | 2 | 334 | 2821759 | ||
BG432594.1 | - | 2 | 387 | |||
BQ073652.1 | - | 2 | 176 | |||
CX786861.1 | - | 2 | 263 |