Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERLIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100626.1 | NP_001094096.1 | 5 | 365 | ||
NM_006459.3 | NP_006450.2 | 4 | 609 | |||
XM_005269442.3 | XP_005269499.1 | 5 | 395 | |||
XM_017015541.1 | XP_016871030.1 | 5 | 379 | |||
GenBank mRNA | AF064093.1 | - | 5 | 341 | ||
AK056574.1 | - | 4 | 2966 | |||
AK222480.1 | - | 5 | 337 | |||
AK298427.1 | - | 4 | 255 | |||
AK315580.1 | - | 5 | 337 | |||
BC031791.1 | - | 4 | 278 | 4214940 | ||
BP280488.1 | - | 5 | 365 | |||
BU192550.1 | - | 4 | 609 |