Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TXNIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006472.5 | NP_006463.3 | 1 | 406 | ||
XM_017000085.1 | XP_016855574.1 | 1 | 355 | |||
GenBank mRNA | AK226090.1 | - | 1 | 318 | ||
AK301203.1 | - | 1 | 354 | |||
AY594328.2 | - | 1 | 41 | |||
BC093702.1 | - | 1 | 115 | 7939547 | ||
BC093704.1 | - | 1 | 115 | 7939549 | ||
BX537824.1 | - | 1 | 354 | |||
DB058019.1 | - | 1 | 382 | |||
HY177853.1 | - | 1 | 408 | |||
S73591.1 | - | 1 | 262 |