Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POSTN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135934.1 | NP_001129406.1 | 17 | 2199 | ||
NM_001135935.1 | NP_001129407.1 | 18 | 2289 | |||
NM_001135936.1 | NP_001129408.1 | 17 | 2199 | |||
NM_001286665.1 | NP_001273594.1 | 18 | 2289 | |||
NM_006475.2 | NP_006466.2 | 19 | 2370 | |||
XM_005266231.2 | XP_005266288.1 | 19 | 2365 | |||
XM_005266232.2 | XP_005266289.1 | 18 | 2275 | |||
XM_017020355.1 | XP_016875844.1 | 19 | 2365 | |||
XM_017020356.1 | XP_016875845.1 | 19 | 2365 | |||
GenBank mRNA | AK021444.1 | - | 1 | 832 | ||
AK297914.1 | - | 8 | 796 | |||
AK304640.1 | - | 8 | 658 | |||
AY140646.1 | - | 18 | 2182 | |||
AY918092.1 | - | 17 | 2092 | |||
BC106709.1 | - | 17 | 2092 | 40007999 | ||
BC106710.1 | - | 17 | 2092 | 40008000 | ||
D13665.1 | - | 17 | 2108 | |||
D13666.1 | - | 19 | 2263 | |||
EU262883.1 | - | 18 | 2171 | |||
EU262885.1 | - | 18 | 2171 | |||
GU354210.1 | - | 18 | 2182 |