Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006635.2 | NP_001006636.1 | 11 | 1172 | ||
NM_001319097.1 | NP_001306026.1 | 10 | 899 | |||
NM_001319098.1 | NP_001306027.1 | 9 | 891 | |||
NM_006554.4 | NP_006545.1 | 10 | 968 | |||
XM_011510497.2 | XP_011508799.1 | 11 | 1065 | |||
GenBank mRNA | AF053551.1 | - | 10 | 941 | ||
AK001287.1 | - | 10 | 920 | |||
AK289359.1 | - | 10 | 919 | |||
AK303564.1 | - | 11 | 1097 | |||
AK315918.1 | - | 11 | 1108 | |||
BC017271.2 | - | 11 | 1102 | 4635884 | ||
BC067831.1 | - | 10 | 947 | 5298149 | ||
BC088359.1 | - | 10 | 903 | 30378040 | ||
CX165048.1 | - | 10 | 792 |