Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CELF2 SIRNA Assays ›
Gene Symbol:
CELF2
Gene Name:
CUGBP, Elav-like family member 2
Gene Aliases:
BRUNOL3, CELF-2, CUG-BP2, CUGBP2, ETR-3, ETR3, NAPOR
Chromosome Location:
Chr.10: 10462515 - 11336675 on Build GRCh38
UniGene ID:
Hs.309288
Species:
Human
siRNA ID
HSS116451
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CELF2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001025076.2
NP_001020247.1
5
685
NM_001025077.2
NP_001020248.1
5
542
NM_001083591.1
NP_001077060.1
5
685
NM_001326317.1
NP_001313246.1
7
544
NM_001326318.1
NP_001313247.1
7
544
NM_001326319.1
NP_001313248.1
8
582
NM_001326320.1
NP_001313249.1
8
638
NM_001326321.1
NP_001313250.1
7
544
NM_001326323.1
NP_001313252.1
8
638
NM_001326324.1
NP_001313253.1
6
521
NM_001326325.1
NP_001313254.1
7
615
NM_001326326.1
NP_001313255.1
6
540
NM_001326327.1
NP_001313256.1
6
540
NM_001326328.1
NP_001313257.1
7
634
NM_001326329.1
NP_001313258.1
5
574
NM_001326330.1
NP_001313259.1
5
574
NM_001326331.1
NP_001313260.1
5
726
NM_001326332.1
NP_001313261.1
5
726
NM_001326333.1
NP_001313262.1
5
726
NM_001326334.1
NP_001313263.1
6
911
NM_001326335.1
NP_001313264.1
5
726
NM_001326336.1
NP_001313265.1
5
726
NM_001326337.1
NP_001313266.1
5
726
NM_001326338.1
NP_001313267.1
4
632
NM_001326339.1
NP_001313268.1
4
632
NM_001326340.1
NP_001313269.1
5
638
NM_001326341.1
NP_001313270.1
5
638
NM_001326342.1
NP_001313271.1
5
638
NM_001326343.1
NP_001313272.1
5
638
NM_001326344.1
NP_001313273.1
5
428
NM_001326345.1
NP_001313274.1
5
506
NM_001326346.1
NP_001313275.1
4
751
NM_001326347.1
NP_001313276.1
5
836
NM_001326348.1
NP_001313277.1
5
834
NM_001326349.1
NP_001313278.1
5
834
NM_006561.3
NP_006552.3
5
631
XM_005252354.4
XP_005252411.1
5
721
XM_005252357.4
XP_005252414.1
4
420
XM_005252358.3
XP_005252415.1
4
761
XM_006717373.2
XP_006717436.1
4
761
XM_011519294.2
XP_011517596.1
5
721
XM_011519297.1
XP_011517599.1
5
844
XM_017015542.1
XP_016871031.1
4
397
XM_017015543.1
XP_016871032.1
5
487
XM_017015544.1
XP_016871033.1
5
487
XM_017015545.1
XP_016871034.1
5
487
XM_017015546.1
XP_016871035.1
5
487
XM_017015547.1
XP_016871036.1
5
721
XM_017015548.1
XP_016871037.1
5
487
XM_017015549.1
XP_016871038.1
5
487
XM_017015550.1
XP_016871039.1
5
1467
XM_017015551.1
XP_016871040.1
5
721
XM_017015552.1
XP_016871041.1
5
844
XM_017015553.1
XP_016871042.1
5
844
XM_017015554.1
XP_016871043.1
5
844
XM_017015555.1
XP_016871044.1
4
397
XM_017015556.1
XP_016871045.1
4
761
XM_017015557.1
XP_016871046.1
6
426
XM_017015558.1
XP_016871047.1
4
413
XM_017015559.1
XP_016871048.1
4
761
XM_017015560.1
XP_016871049.1
4
422
XM_017015561.1
XP_016871050.1
4
749
XM_017015562.1
XP_016871051.1
4
761
XM_017015563.1
XP_016871052.1
4
748
XM_017015564.1
XP_016871053.1
4
417
XM_017015565.1
XP_016871054.1
4
420
XM_017015566.1
XP_016871055.1
4
417
XM_017015567.1
XP_016871056.1
5
557
XM_017015568.1
XP_016871057.1
7
544
XM_017015569.1
XP_016871058.1
5
844
XM_017015570.1
XP_016871059.1
4
420
XM_017015571.1
XP_016871060.1
5
844
XM_017015572.1
XP_016871061.1
5
545
GenBank mRNA
AF036956.1
-
5
601
AF090693.1
-
5
517
AF090694.1
-
5
469
AF432906.1
-
5
537
AK293164.1
-
5
578
AK295511.1
-
3
272
AK297313.1
-
4
542
AK297380.1
-
4
420
AK299548.1
-
5
461
AK299983.1
-
5
750
AK299992.1
-
5
618
AK302681.1
-
3
316
AK304216.1
-
4
427
AK310038.1
-
3
316
AK314087.1
-
5
640
AK316354.1
-
5
437
BC036391.1
-
5
603
5260623
BP362205.1
-
3
184
BQ213648.1
-
5
686
DA819821.1
-
4
355
U69546.1
-
5
418
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
RNA processing
mRNA processing
regulation of heart contraction
nucleotide binding
RNA binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-wjdsd:80/100.66.76.85:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline