Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C6orf10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286474.1 | NP_001273403.1 | 26 | 1606 | ||
NM_001286475.1 | NP_001273404.1 | 24 | 1564 | |||
NM_006781.4 | NP_006772.3 | 23 | 1612 | |||
XM_017010182.1 | XP_016865671.1 | 16 | 1133 | |||
XM_017010185.1 | XP_016865674.1 | 24 | 1637 | |||
GenBank mRNA | AK225742.1 | - | 26 | 1597 | ||
AK302176.1 | - | 24 | 1524 | |||
BC034774.1 | - | 23 | 1579 | 5169157 | ||
BX113229.1 | - | 1 | 152 | |||
U60665.1 | - | 26 | 1633 |