Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CGRRF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006568.2 | NP_006559.1 | 6 | 1015 | ||
XM_011536362.2 | XP_011534664.1 | 6 | 904 | |||
XM_017020931.1 | XP_016876420.1 | 7 | 1106 | |||
XM_017020932.1 | XP_016876421.1 | 7 | 972 | |||
GenBank mRNA | AK090727.1 | - | 2 | 2004 | ||
AK289478.1 | - | 6 | 1046 | |||
AW664053.1 | - | 1 | 310 | |||
BC015063.1 | - | 6 | 981 | 4245551 | ||
U66469.1 | - | 6 | 951 |