Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006579.2 | NP_006570.1 | 2 | 455 | ||
GenBank mRNA | BC001549.1 | - | 2 | 385 | 3463689 | |
BC001572.1 | - | 2 | 387 | 3453423 | ||
BC046501.1 | - | 2 | 429 | 5106272 | ||
CA488777.1 | - | 2 | 435 | |||
CR456815.1 | - | 1 | 250 | |||
CR542094.1 | - | 1 | 250 | |||
KJ921656.1 | - | 1 | 250 | |||
Z37986.1 | - | 2 | 361 |