Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GMEB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319674.1 | NP_001306603.1 | 9, 10 | 1123 | ||
NM_006582.3 | NP_006573.2 | 9, 10 | 1153 | |||
NM_024482.2 | NP_077808.1 | 9, 10 | 1120 | |||
XM_011540518.2 | XP_011538820.1 | 9, 10 | 1256 | |||
XM_011540519.2 | XP_011538821.1 | 9, 10 | 1096 | |||
XM_011540521.2 | XP_011538823.1 | 9, 10 | 1225 | |||
XM_011540522.2 | XP_011538824.1 | 9, 10 | 1072 | |||
XM_017000087.1 | XP_016855576.1 | 9, 10 | 1271 | |||
GenBank mRNA | AF099013.1 | - | 8, 9 | 1017 | ||
AF173868.1 | - | 9, 10 | 1048 | |||
AF203694.1 | - | 9, 10 | 1047 | |||
AK000892.1 | - | 9, 10 | 1119 | |||
AK023126.1 | - | 9, 10 | 1040 | |||
AK315195.1 | - | 9, 10 | 1099 | |||
AK315436.1 | - | 9, 10 | 1057 | |||
BC001473.2 | - | 9, 10 | 1070 | 3138481 |