Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CORIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278586.1 | NP_001265515.1 | 3 | 432 | ||
NM_006587.3 | NP_006578.2 | 3 | 432 | |||
GenBank mRNA | AF133845.1 | - | 3 | 368 | ||
AK297675.1 | - | 3 | 393 | |||
AK302816.1 | - | 2 | 107 | |||
AK304466.1 | - | 3 | 432 | |||
BC110451.2 | - | 3 | 368 | 40027293 | ||
BX648285.1 | - | 3 | 433 |