Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006593.3 | NP_006584.1 | 2, 3 | 1163 | ||
GenBank mRNA | AK226050.1 | - | 1, 2 | 1499 | ||
AK299533.1 | - | 1, 2 | 63 | |||
AK312567.1 | - | 2, 3 | 938 | |||
BC029289.1 | - | 2, 3 | 924 | |||
BC104844.1 | - | 2, 3 | 1091 | 8143847 | ||
BC113418.1 | - | 2, 3 | 1091 | 8322470 | ||
U49250.1 | - | 2, 3 | 870 |