Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC12A7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006598.2 | NP_006589.2 | 17, 18 | 2310 | ||
XM_005248231.3 | XP_005248288.1 | 17, 18 | 2315 | |||
XM_011513939.2 | XP_011512241.1 | 17, 18 | 2404 | |||
XM_011513940.2 | XP_011512242.1 | 17, 18 | 2239 | |||
XM_011513941.2 | XP_011512243.2 | 17, 18 | 2334 | |||
XM_017008958.1 | XP_016864447.1 | 17, 18 | 2614 | |||
XM_017008959.1 | XP_016864448.1 | 17, 18 | 2266 | |||
GenBank mRNA | AF105365.1 | - | 17, 18 | 2248 | ||
AK024493.1 | - | 10, 11 | 1335 | |||
AK024497.1 | - | 10, 11 | 1160 | |||
BC098390.1 | - | 17, 18 | 2288 | 5265743 |