Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFAT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113178.2 | NP_001106649.1 | 13 | 2410 | ||
NM_006599.3 | NP_006590.1 | 12 | 2359 | |||
NM_138713.3 | NP_619727.2 | 13 | 2413 | |||
NM_138714.3 | NP_619728.2 | 14 | 2475 | |||
NM_173214.2 | NP_775321.1 | 13 | 2421 | |||
NM_173215.2 | NP_775322.1 | 13 | 2421 | |||
XM_006721125.3 | XP_006721188.1 | 12 | 3084 | |||
XM_011522817.2 | XP_011521119.1 | 12 | 2356 | |||
XM_011522818.2 | XP_011521120.1 | 15 | 2548 | |||
XM_011522819.2 | XP_011521121.1 | 15 | 2130 | |||
XM_011522820.2 | XP_011521122.1 | 12 | 1560 | |||
XM_011522822.2 | XP_011521124.1 | 11 | 1508 | |||
XM_017022870.1 | XP_016878359.1 | 12 | 2708 | |||
XM_017022871.1 | XP_016878360.1 | 13 | 2432 | |||
XM_017022872.1 | XP_016878361.1 | 12 | 2289 | |||
XM_017022873.1 | XP_016878362.1 | 12 | 1898 | |||
GenBank mRNA | AB020634.1 | - | 12 | 2341 | ||
AF089824.1 | - | 13 | 2363 | |||
AF134870.1 | - | 10 | 1795 | |||
AF163836.1 | - | 11 | 1882 | |||
AF346509.1 | - | 14 | 3361 | |||
AJ005683.1 | - | 13 | 2037 | |||
AJ005684.1 | - | 13 | 2037 | |||
AJ243298.1 | - | 13 | 2180 | |||
AJ243299.1 | - | 13 | 2172 | |||
BC131509.1 | - | 13 | 2314 | 40083218 | ||
BC146765.1 | - | 12 | 2341 | 9007208 |