Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFAT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113178.2 | NP_001106649.1 | 13 | 2863 | ||
NM_006599.3 | NP_006590.1 | 12 | 2812 | |||
NM_138713.3 | NP_619727.2 | 13 | 2866 | |||
NM_138714.3 | NP_619728.2 | 14 | 2928 | |||
NM_173214.2 | NP_775321.1 | 13 | 2874 | |||
NM_173215.2 | NP_775322.1 | 13 | 2874 | |||
XM_006721125.3 | XP_006721188.1 | 12 | 3537 | |||
XM_011522817.2 | XP_011521119.1 | 12 | 2809 | |||
XM_011522818.2 | XP_011521120.1 | 15 | 3001 | |||
XM_011522819.2 | XP_011521121.1 | 15 | 2583 | |||
XM_011522820.2 | XP_011521122.1 | 12 | 2013 | |||
XM_011522822.2 | XP_011521124.1 | 11 | 1961 | |||
XM_017022870.1 | XP_016878359.1 | 12 | 3161 | |||
XM_017022871.1 | XP_016878360.1 | 13 | 2885 | |||
XM_017022872.1 | XP_016878361.1 | 12 | 2742 | |||
XM_017022873.1 | XP_016878362.1 | 12 | 2351 | |||
GenBank mRNA | AB020634.1 | - | 12 | 2794 | ||
AF089824.1 | - | 13 | 2816 | |||
AF134870.1 | - | 10 | 2248 | |||
AF163836.1 | - | 11 | 2335 | |||
AF346509.1 | - | 14 | 3814 | |||
AJ005683.1 | - | 13 | 2490 | |||
AJ005684.1 | - | 13 | 2490 | |||
AJ243298.1 | - | 13 | 2633 | |||
AJ243299.1 | - | 13 | 2625 | |||
BC131509.1 | - | 13 | 2767 | 40083218 | ||
BC146765.1 | - | 12 | 2794 | 9007208 | ||
Z97016.1 | - | 1 | 455 |