Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YME1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253866.1 | NP_001240795.1 | 11, 12 | 1403 | ||
NM_014263.3 | NP_055078.1 | 12, 13 | 1502 | |||
NM_139312.2 | NP_647473.1 | 13, 14 | 1673 | |||
XM_011519300.2 | XP_011517602.1 | 12, 13 | 1956 | |||
GenBank mRNA | AF070656.1 | - | 8, 9 | 817 | ||
AF151782.1 | - | 13, 14 | 1517 | |||
AJ132637.1 | - | 12, 13 | 1475 | |||
AK001259.1 | - | 3, 4 | 264 | |||
AK291292.1 | - | 12, 13 | 1431 | |||
AK297973.1 | - | 11, 12 | 1367 | |||
AK309708.1 | - | 8, 9 | 948 | |||
AK314372.1 | - | 12, 13 | 1430 | |||
AY358484.1 | - | 12, 13 | 1428 | |||
BC007795.2 | - | 13, 14 | 1609 | 4110395 | ||
BC023507.2 | - | 12, 13 | 1444 | 3637576 | ||
BC024032.2 | - | 12, 13 | 1449 | 2961446 |