Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YME1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253866.1 | NP_001240795.1 | 3 | 405 | ||
NM_014263.3 | NP_055078.1 | 3 | 405 | |||
NM_139312.2 | NP_647473.1 | 4 | 576 | |||
XM_011519300.2 | XP_011517602.1 | 4 | 958 | |||
GenBank mRNA | AJ132637.1 | - | 3 | 378 | ||
AK291292.1 | - | 3 | 334 | |||
AK297973.1 | - | 3 | 369 | |||
AK314372.1 | - | 3 | 333 | |||
AY358484.1 | - | 3 | 331 | |||
BC007795.2 | - | 4 | 512 | 4110395 | ||
BC019602.1 | - | 3 | 2391 | |||
BC023507.2 | - | 3 | 347 | 3637576 | ||
BC024032.2 | - | 3 | 352 | 2961446 | ||
DA489399.1 | - | 4 | 537 |