Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAF3IP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164281.2 | NP_001157753.1 | 2 | 968 | ||
NM_147200.2 | NP_671733.2 | 3 | 756 | |||
NM_147686.3 | NP_679211.2 | 2 | 968 | |||
XM_011535386.1 | XP_011533688.1 | 3 | 1691 | |||
GenBank mRNA | AF136405.1 | - | 3 | 664 | ||
AF136406.1 | - | 2 | 755 | |||
AF136407.1 | - | 2 | 755 | |||
AF272151.1 | - | 1 | 488 | |||
AF274303.1 | - | 2 | 488 | |||
AK025351.1 | - | 2 | 565 | |||
AK310141.1 | - | 2 | 659 | |||
AK310922.1 | - | 3 | 687 | |||
AK314415.1 | - | 3 | 679 | |||
AL050289.1 | - | 2 | 640 | |||
BC002823.2 | - | 2 | 551 | 3634505 |