Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016272.3 | NP_057356.1 | 2 | 739 | ||
XM_005261315.2 | XP_005261372.1 | 2 | 1950 | |||
XM_006724105.3 | XP_006724168.1 | 2 | 1256 | |||
XM_017028539.1 | XP_016884028.1 | 2 | 3032 | |||
GenBank mRNA | AB035207.1 | - | 1 | 246 | ||
AB051450.1 | - | 2 | 498 | |||
BC028919.1 | - | 2 | 488 | 4940977 | ||
BC038957.1 | - | 2 | 512 | 5786640 | ||
CR456594.1 | - | 2 | 192 | |||
CR541684.1 | - | 1 | 32 | |||
D64109.1 | - | 1 | 125 |