Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HBS1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145158.1 | NP_001138630.1 | 6 | 967 | ||
NM_006620.3 | NP_006611.1 | 7 | 1093 | |||
GenBank mRNA | AB028961.1 | - | 2 | 322 | ||
AJ420551.1 | - | 7 | 1010 | |||
AJ459826.1 | - | 7 | 1079 | |||
AK024258.1 | - | 7 | 1014 | |||
AK293573.1 | - | 4 | 301 | |||
AK295545.1 | - | 6 | 888 | |||
AK298336.1 | - | 4 | 496 | |||
BC001465.2 | - | 7 | 1002 | 3138910 | ||
BC040849.1 | - | 7 | 997 | 5104748 | ||
BX648033.1 | - | 4 | 758 | |||
U87791.1 | - | 7 | 1080 |