Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252226.1 | NP_001239155.1 | 2, 3 | 403 | ||
NM_006622.3 | NP_006613.2 | 1, 2 | 445 | |||
GenBank mRNA | AF059617.1 | - | 1, 2 | 565 | ||
AF223574.1 | - | 1, 2 | 388 | |||
AK027851.1 | - | 1, 2 | 390 | |||
AK098163.1 | - | 2, 3 | 371 | |||
AK297298.1 | - | 2, 3 | 348 | |||
AK312505.1 | - | 1, 2 | 393 | |||
BC013879.2 | - | 1, 2 | 378 | 3831747 | ||
BI559351.1 | - | 449 | ||||
DA020501.1 | - | 1, 2 | 392 |