Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006627.2 | NP_006618.1 | 7 | 619 | ||
NR_027368.1 | 6 | 395 | ||||
GenBank mRNA | AF001176.2 | - | 7 | 590 | ||
AK303791.1 | - | 6 | 361 | |||
AK314790.1 | - | 7 | 612 | |||
BC004438.1 | - | 7 | 579 | 2820501 | ||
BC006098.1 | - | 7 | 585 | 2989197 | ||
CR536569.1 | - | 6 | 563 | |||
CR936714.1 | - | 6 | 365 | |||
Y18863.1 | - | 7 | 588 |