Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEK6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145001.2 | NP_001138473.1 | 7 | 808 | ||
NM_001166167.1 | NP_001159639.1 | 6 | 590 | |||
NM_001166168.1 | NP_001159640.1 | 6 | 585 | |||
NM_001166169.1 | NP_001159641.1 | 6 | 594 | |||
NM_001166170.1 | NP_001159642.1 | 6 | 592 | |||
NM_001166171.1 | NP_001159643.1 | 7 | 757 | |||
NM_014397.5 | NP_055212.2 | 6 | 643 | |||
XM_005251664.1 | XP_005251721.1 | 6 | 649 | |||
XM_006716936.2 | XP_006716999.1 | 6 | 654 | |||
XM_017014217.1 | XP_016869706.1 | 7 | 817 | |||
GenBank mRNA | AB026289.1 | - | 6 | 574 | ||
AF087909.1 | - | 6 | 579 | |||
AK294614.1 | - | 6 | 594 | |||
AK313071.1 | - | 6 | 544 | |||
BC000101.2 | - | 6 | 589 | 3507262 | ||
BC004174.2 | - | 7 | 761 | 2958695 | ||
BC004209.2 | - | 7 | 761 | 2958695 | ||
BC012761.2 | - | 6 | 590 | 3640103 | ||
CR457091.1 | - | 5 | 424 | |||
CR542222.1 | - | 5 | 424 |