Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTHFD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006636.3 | NP_006627.2 | 8 | 1092 | ||
NR_027405.1 | 9 | 1190 | ||||
XM_006711924.2 | XP_006711987.1 | 7 | 879 | |||
GenBank mRNA | AK130664.1 | - | 8 | 1040 | ||
AK222826.1 | - | 8 | 1042 | |||
AK223041.1 | - | 8 | 1040 | |||
AK300035.1 | - | 7 | 857 | |||
AK302245.1 | - | 6 | 732 | |||
AK313138.1 | - | 8 | 1040 | |||
BC001548.1 | - | 8 | 1027 | 3462450 | ||
BC015062.1 | - | 9 | 1140 | 4285669 | ||
BC017054.1 | - | 8 | 1020 | 3846710 | ||
X16396.1 | - | 8 | 1010 |