Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPLX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006651.3 | NP_006642.1 | 2 | 204 | ||
GenBank mRNA | AB593095.1 | - | 2 | 181 | ||
AF022383.1 | - | 1 | 121 | |||
AK126578.1 | - | 2 | 139 | |||
AK311921.1 | - | 2 | 176 | |||
BC002471.2 | - | 2 | 204 | 3349779 | ||
BT007029.1 | - | 1 | 13 | |||
KJ534815.1 | - | 1 | 13 |