Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPLX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006651.3 | NP_006642.1 | 3, 4 | 395 | ||
XM_011513391.1 | XP_011511693.1 | 2, 3 | 326 | |||
GenBank mRNA | AB593095.1 | - | 3, 4 | 372 | ||
AF022383.1 | - | 2, 3 | 312 | |||
AK126578.1 | - | 4, 5 | 267 | |||
AK311921.1 | - | 3, 4 | 367 | |||
BC002471.2 | - | 3, 4 | 395 | 3349779 | ||
BT007029.1 | - | 2, 3 | 204 | |||
KJ534815.1 | - | 2, 3 | 204 |