Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPLX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006651.3 | NP_006642.1 | 4 | 582 | ||
XM_011513391.1 | XP_011511693.1 | 3 | 513 | |||
GenBank mRNA | AB593095.1 | - | 4 | 559 | ||
AF022383.1 | - | 3 | 499 | |||
AK126578.1 | - | 5 | 454 | |||
AK311921.1 | - | 4 | 554 | |||
BC002471.2 | - | 4 | 582 | 3349779 | ||
BT007029.1 | - | 3 | 391 |