Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDH1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270364.1 | NP_001257293.1 | 21 | 2632 | ||
NM_001270365.1 | NP_001257294.1 | 19 | 2431 | |||
NM_012190.3 | NP_036322.2 | 21 | 2734 | |||
NR_072979.1 | 19 | 2483 | ||||
XM_006713481.2 | XP_006713544.1 | 21 | 2473 | |||
XM_011512355.1 | XP_011510657.1 | 21 | 2466 | |||
XM_017005613.1 | XP_016861102.1 | 19 | 2635 | |||
XM_017005614.1 | XP_016861103.1 | 19 | 2251 | |||
GenBank mRNA | AF052732.1 | - | 21 | 2494 | ||
AK124908.1 | - | 4 | 1249 | |||
AK222535.1 | - | 21 | 2516 | |||
AK222694.1 | - | 21 | 2516 | |||
AK294392.1 | - | 19 | 2215 | |||
BC027241.1 | - | 19 | 2256 | 4484301 | ||
CR749807.1 | - | 21 | 2632 |