Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDH1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270364.1 | NP_001257293.1 | 21 | 2679 | ||
NM_001270365.1 | NP_001257294.1 | 19 | 2478 | |||
NM_012190.3 | NP_036322.2 | 21 | 2781 | |||
NR_072979.1 | 19 | 2530 | ||||
XM_006713481.2 | XP_006713544.1 | 21 | 2520 | |||
XM_011512355.1 | XP_011510657.1 | 21 | 2513 | |||
XM_017005613.1 | XP_016861102.1 | 19 | 2682 | |||
XM_017005614.1 | XP_016861103.1 | 19 | 2298 | |||
GenBank mRNA | AA639766.1 | - | 3 | 518 | ||
AB209299.1 | - | 21 | 2588 | |||
AF052732.1 | - | 21 | 2541 | |||
AK124908.1 | - | 4 | 1296 | |||
AK222535.1 | - | 21 | 2563 | |||
AK222694.1 | - | 21 | 2563 | |||
AK294392.1 | - | 19 | 2262 | |||
BC027241.1 | - | 19 | 2303 | 4484301 | ||
CR749807.1 | - | 21 | 2679 |